Assay Details
Target Gene Details
Entrez Gene ID: | 6492 |
Gene Name: | single-minded family bHLH transcription factor 1 |
Gene Aliases: |
bHLHe14 |
Location: |
Chr.6:100385009-100464929 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIM1 | NM_005068.2 | 10 | 1515 | NP_005059.2 |
| XM_005267100.2 | 11 | 2091 | XP_005267157.1 | |
| XM_011536072.2 | 11 | 2091 | XP_011534374.1 | |
| XM_017011197.1 | 11 | 1823 | XP_016866686.1 | |
| AK308090.1 | 11 | 2091 | ||
| U70212.1 | 10 | 1524 | AAB62395.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1028122 | Chr.6:100326369 - 100516471 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map