Assay Details
Target Gene Details
Entrez Gene ID: | 9637 |
Gene Name: | fasciculation and elongation protein zeta 2 |
Gene Aliases: |
HUM3CL |
Location: |
Chr.2:36552239-36598190 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FEZ2 | NM_001042548.1 | NP_001036013.1 | ||
| NM_005102.2 | NP_005093.2 | |||
| XM_006712153.2 | XP_006712216.1 | |||
| AB076184.1 | BAD06207.1 | |||
| AB209719.1 | BAD92956.1 | |||
| AF113124.1 | AAF14865.1 | |||
| AK025274.1 | ||||
| BC089390.1 | AAH89390.1 | |||
| BC110386.1 | ||||
| BX537785.1 | CAD97842.1 | |||
| U60061.1 | AAC51283.1 | |||
| U69140.1 | AAB40661.1 |
More Information
Set Membership: |
|

Genomic Map