Assay Details
Target Gene Details
Entrez Gene ID: | 374378 |
Gene Name: | polypeptide N-acetylgalactosaminyltransferase 18 |
Gene Aliases: |
GALNACT18, GALNT15, GALNTL4, GalNAc-T15, GalNAc-T18 |
Location: |
Chr.11:11270874-11622045 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALNT18 | NM_198516.2 | 11 | 2215 | NP_940918.2 |
| XM_006718224.2 | 10 | 2060 | XP_006718287.1 | |
| XM_011520068.2 | 11 | 1791 | XP_011518370.1 | |
| AB078147.1 | BAD93179.1 | |||
| AJ626724.1 | CAF25035.1 | |||
| AK055111.1 | 11 | 2197 | ||
| AK124690.1 | 1 | 3514 | ||
| BC037341.2 | 11 | 2204 | AAH37341.3 | |
| BC060864.1 | 11 | 2206 | AAH60864.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832064 | Chr.11:11128217 - 11316356 on Build GRCh38 | Gain |
|
| nsv483000 | Chr.11:10678454 - 12678453 on Build GRCh38 | Loss |
|
| nsv509389 | Chr.11:11214235 - 11286237 on Build GRCh38 | Insertion |
|
| esv34098 | Chr.11:11248654 - 11450079 on Build GRCh38 | Loss |
|
| esv32920 | Chr.11:11194328 - 11359745 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map