Assay Details
Target Gene Details
Entrez Gene ID: | 80258 |
Gene Name: | EF-hand domain containing 2 |
Gene Aliases: |
MRX74, dJ1158H2.1 |
Location: |
Chr.X:44146254-44343677 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EFHC2 | NM_025184.3 | 15 | 2328 | NP_079460.2 |
| XM_006724562.2 | 14 | 2041 | XP_006724625.1 | |
| AK026254.1 | 5 | 602 | BAB15413.1 | |
| AK026496.1 | 15 | 2180 | BAB15492.1 | |
| AK291187.1 | 15 | 2469 | ||
| AK292967.1 | 15 | 2312 | ||
| BC031039.1 | 15 | 2323 | AAH31039.1 | |
| CR749363.1 | 10 | 1991 | CAH18216.1 |
More Information
Set Membership: |
|

Genomic Map