Assay Details
Target Gene Details
Entrez Gene ID: | 25896 |
Gene Name: | integrator complex subunit 7 |
Gene Aliases: |
C1orf73, INT7 |
Location: |
Chr.1:211940328-212035660 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INTS7 | NM_001199809.1 | 19 | 2917 | NP_001186738.1 |
| NM_001199811.1 | 20 | 3022 | NP_001186740.1 | |
| NM_001199812.1 | 20 | 3004 | NP_001186741.1 | |
| NM_015434.3 | 20 | 3064 | NP_056249.1 | |
| NR_037667.1 | 21 | 3098 | ||
| XM_011509396.2 | 21 | 3057 | XP_011507698.1 | |
| AK001363.1 | 20 | 2864 | BAA91650.1 | |
| AK001598.1 | 20 | 2946 | BAA91779.1 | |
| AK022509.1 | 20 | 2924 | BAB14067.1 | |
| AK022589.1 | 12 | 1731 | BAB14116.1 | |
| AK297225.1 | 19 | 2796 | ||
| AL117576.1 | 9 | 1338 | CAB56000.1 | |
| AL133049.2 | 15 | 2233 | CAB61376.1 | |
| BC020523.2 | 20 | 2898 | AAH20523.1 | |
| BC030716.1 | 20 | 2919 | AAH30716.1 | |
| BC033918.1 | 20 | 2923 | AAH33918.1 |
More Information
Set Membership: |
|

Genomic Map