Assay Details
Target Gene Details
Entrez Gene ID: | 27107 |
Gene Name: | zinc finger and BTB domain containing 11 |
Gene Aliases: |
ZNF-U69274, ZNF913 |
Location: |
Chr.3:101649439-101677144 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZBTB11 | NM_014415.3 | NP_055230.2 | ||
| AK312859.1 | ||||
| BC111700.1 | AAI11701.1 | |||
| DA907277.1 | ||||
| U69274.1 | AAD00172.1 |
Target Gene Details
Entrez Gene ID: | 100009676 |
Gene Name: | ZBTB11 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:101676430-101679217 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZBTB11-AS1 | NR_024407.1 | 1 | 187 | |
| AK022880.1 | 1 | 171 |
More Information
Set Membership: |
|

Genomic Map