Assay Details
Target Gene Details
Entrez Gene ID: | 149483 |
Gene Name: | coiled-coil domain containing 17 |
Gene Aliases: |
- |
Location: |
Chr.1:45620044-45624787 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC17 | NM_001114938.2 | NP_001108410.2 | ||
| NM_001190182.1 | NP_001177111.1 | |||
| XM_011540812.1 | XP_011539114.1 | |||
| XM_011540814.1 | XP_011539116.1 | |||
| XM_017000450.1 | XP_016855939.1 | |||
| XM_017000451.1 | XP_016855940.1 | |||
| XM_017000452.1 | XP_016855941.1 | |||
| XM_017000453.1 | XP_016855942.1 | |||
| XM_017000454.1 | XP_016855943.1 | |||
| XM_017000455.1 | XP_016855944.1 | |||
| XM_017000456.1 | XP_016855945.1 | |||
| AK057646.1 | 11 | 1738 | BAB71539.1 | |
| AK298104.1 | ||||
| AK303939.1 | ||||
| BC128178.1 | ||||
| BU677500.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893512 | Chr.1:45476180 - 46068871 on Build GRCh38 | Gain |
|
| esv3585911 | Chr.1:45616829 - 45630555 on Build GRCh38 | Gain |
|
| nsv822509 | Chr.1:45619307 - 45623970 on Build GRCh38 | Loss |
|
| nsv829726 | Chr.1:45431909 - 45637514 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map