Assay Details
Target Gene Details
Entrez Gene ID: | 283687 |
Gene Name: | ST20 antisense RNA 1 |
Gene Aliases: |
C15orf37 |
Location: |
Chr.15:79922771-79924854 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ST20-AS1 | NR_028330.1 | 1 | 1320 | |
| AK091107.1 | 1 | 1320 | BAC03584.1 | |
| AK092679.1 | 1 | 926 | ||
| BC016975.1 | 1 | 914 | AAH16975.1 | |
| BG677098.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833066 | Chr.15:79878078 - 80033241 on Build GRCh38 | Loss |
|
| esv3637009 | Chr.15:79898247 - 79960449 on Build GRCh38 | Loss |
|
| nsv833065 | Chr.15:79760661 - 79982480 on Build GRCh38 | Loss |
|
| esv2660680 | Chr.15:79898180 - 79960294 on Build GRCh38 | Deletion |
|
| esv3581779 | Chr.15:79902138 - 79936035 on Build GRCh38 | Loss |
|

Genomic Map