Assay Details
Target Gene Details
Entrez Gene ID: | 55846 |
Gene Name: | integrin alpha FG-GAP repeat containing 2 |
Gene Aliases: |
MDS028 |
Location: |
Chr.12:2812621-2832217 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITFG2 | NR_130744.1 | |||
| AK300905.1 |
Target Gene Details
Entrez Gene ID: | 83714 |
Gene Name: | nuclear receptor interacting protein 2 |
Gene Aliases: |
- |
Location: |
Chr.12:2825348-2835536 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NRIP2 | NM_031474.2 | 6 | 1147 | NP_113662.1 |
| XM_005253797.3 | 7 | 1159 | XP_005253854.1 | |
| XM_005253798.3 | 6 | 1055 | XP_005253855.1 | |
| XM_011521031.2 | 7 | 1089 | XP_011519333.1 | |
| XM_017020000.1 | 4 | 1720 | XP_016875489.1 | |
| XM_017020001.1 | 7 | 846 | XP_016875490.1 | |
| AK054740.1 | 2 | 358 | ||
| AK098063.1 | 6 | 1118 | ||
| AK300944.1 | 8 | 944 | ||
| AL136557.1 | 6 | 1115 | CAB66492.1 | |
| BC036063.1 | 6 | 1116 | AAH36063.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3628321 | Chr.12:2796341 - 2902786 on Build GRCh38 | Gain |
|
| esv3628322 | Chr.12:2796908 - 2837171 on Build GRCh38 | Gain |
|
| dgv19n68 | Chr.12:2726070 - 2935438 on Build GRCh38 | Loss |
|
| nsv832311 | Chr.12:2810344 - 2996539 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map