Assay Details
Target Gene Details
Entrez Gene ID: | 2268 |
Gene Name: | FGR proto-oncogene, Src family tyrosine kinase |
Gene Aliases: |
SRC2, c-fgr, c-src2, p55-Fgr, p55c-fgr, p58-Fgr, p58c-fgr |
Location: |
Chr.1:27612289-27635561 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FGR | NM_001042729.1 | NP_001036194.1 | ||
| NM_001042747.1 | NP_001036212.1 | |||
| NM_005248.2 | NP_005239.1 | |||
| XM_006710452.2 | XP_006710515.1 | |||
| XM_011541010.1 | XP_011539312.1 | |||
| XM_011541011.2 | XP_011539313.1 | |||
| XM_011541012.1 | XP_011539314.1 | |||
| XM_011541013.2 | XP_011539315.1 | |||
| XM_017000673.1 | XP_016856162.1 | |||
| XM_017000674.1 | XP_016856163.1 | |||
| AK125143.1 | ||||
| AK130188.1 | ||||
| AK301078.1 | ||||
| AK304846.1 | ||||
| BC064382.1 | AAH64382.1 | |||
| BI771758.1 | ||||
| BX495937.1 | ||||
| DA139476.1 | ||||
| DA952771.1 | ||||
| DA960178.1 | ||||
| M19722.1 | 1 | 144 | AAA52451.1 | |
| M27454.1 | AAA35679.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508700 | Chr.1:27556643 - 27653685 on Build GRCh38 | Deletion |
|
| dgv220n54 | Chr.1:27522789 - 27631734 on Build GRCh38 | Loss |
|
| nsv1014991 | Chr.1:27560502 - 27628874 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map