Assay Details
Target Gene Details
Entrez Gene ID: | 2595 |
Gene Name: | glucosidase alpha, neutral C |
Gene Aliases: |
- |
Location: |
Chr.15:42273658-42353666 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GANC | NM_001301409.1 | 11 | 1656 | NP_001288338.1 |
| NM_198141.2 | 10 | 1269 | NP_937784.2 | |
| AF525397.1 | 6 | 522 | AAO14993.1 | |
| AF545045.1 | 10 | 963 | AAN74756.1 | |
| AF545046.1 | 10 | 963 | AAN74757.1 | |
| AK074037.1 | 10 | 1161 | ||
| AK074330.1 | 11 | 1658 | ||
| BC059406.1 | 10 | 1141 | AAH59406.1 | |
| BC093833.1 | 10 | 986 | AAH93833.1 | |
| BC112051.1 | 10 | 986 | AAI12052.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3636263 | Chr.15:42288763 - 42328316 on Build GRCh38 | Loss |
|
| nsv832986 | Chr.15:42295627 - 42461466 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map