Assay Details
Target Gene Details
Entrez Gene ID: | 255394 |
Gene Name: | t-complex 11 like 2 |
Gene Aliases: |
- |
Location: |
Chr.12:106302026-106347455 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TCP11L2 | NM_001286262.1 | NP_001273191.1 | ||
| NM_152772.2 | NP_689985.1 | |||
| XM_005268767.4 | XP_005268824.1 | |||
| XM_005268768.4 | XP_005268825.1 | |||
| XM_011538129.2 | XP_011536431.1 | |||
| XM_011538130.2 | XP_011536432.1 | |||
| XM_011538131.2 | XP_011536433.1 | |||
| XM_011538132.2 | XP_011536434.1 | |||
| XM_017019127.1 | XP_016874616.1 | |||
| XM_017019128.1 | XP_016874617.1 | |||
| XM_017019129.1 | XP_016874618.1 | |||
| XM_017019130.1 | XP_016874619.1 | |||
| XM_017019131.1 | XP_016874620.1 | |||
| XM_017019132.1 | XP_016874621.1 | |||
| XM_017019133.1 | XP_016874622.1 | |||
| AF306671.1 | ||||
| AF306672.1 | ||||
| AF306673.1 | ||||
| AF306674.1 | ||||
| AF306858.1 | ||||
| AK314055.1 | ||||
| BC033617.1 | AAH33617.1 |
More Information
Set Membership: |
|

Genomic Map