Assay Details
Target Gene Details
Entrez Gene ID: | 7050 |
Gene Name: | TGFB induced factor homeobox 1 |
Gene Aliases: |
HPE4, TGIF |
Location: |
Chr.18:3411927-3458412 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
More Information
Set Membership: |
|

Genomic Map