Assay Details
Target Gene Details
Entrez Gene ID: | 285313 |
Gene Name: | immunoglobulin superfamily member 10 |
Gene Aliases: |
CMF608 |
Location: |
Chr.3:151435051-151619926 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IGSF10 | NM_001178145.1 | 2 | 1316 | NP_001171616.1 |
| NM_001178146.1 | 2 | 1163 | NP_001171617.1 | |
| NM_178822.4 | 6 | 7063 | NP_849144.2 | |
| XM_011512708.2 | 12 | 7872 | XP_011511010.1 | |
| XM_011512709.2 | 11 | 7603 | XP_011511011.1 | |
| AK093069.1 | 2 | 1299 | BAC04042.1 | |
| AK098838.1 | 1 | 506 | BAC05429.1 | |
| AK128160.1 | 3 | 3113 | BAC87305.1 | |
| AY273815.1 | 6 | 7063 | AAQ16156.1 | |
| BC031063.1 | 2 | 1153 | AAH31063.1 | |
| BC045734.1 | 2 | 1153 | AAH45734.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829757 | Chr.3:151327092 - 151508654 on Build GRCh38 | Loss |
|
| nsv4063 | Chr.3:151408925 - 151444402 on Build GRCh38 | Insertion |
|
| nsv508960 | Chr.3:151425271 - 151443978 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map