Assay Details
Target Gene Details
Entrez Gene ID: | 5451 |
Gene Name: | POU class 2 homeobox 1 |
Gene Aliases: |
OCT1, OTF1, oct-1B |
Location: |
Chr.1:167220829-167427345 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POU2F1 | NM_001198786.1 | 1 | 92 | NP_001185715.1 |
| NM_002697.3 | 1 | 92 | NP_002688.3 | |
| NR_037163.1 | ||||
| XM_011509653.2 | XP_011507955.1 | |||
| XM_011509655.1 | XP_011507957.1 | |||
| XM_017001508.1 | XP_016856997.1 | |||
| AK091438.1 | ||||
| AK302525.1 | ||||
| AK303201.1 | ||||
| AK308004.1 | ||||
| BC052274.1 | AAH52274.1 | |||
| BX954888.1 | 1 | 93 | ||
| DC370634.1 | ||||
| S66901.1 | AAB28879.1 | |||
| X13403.1 | CAA31767.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521232 | Chr.1:167191722 - 167388324 on Build GRCh38 | Gain+Loss |
|
| nsv1078309 | Chr.1:167219462 - 167223763 on Build GRCh38 | Deletion |
|
| nsv3421 | Chr.1:167194524 - 167224129 on Build GRCh38 | Insertion |
|
| nsv831803 | Chr.1:167215480 - 167396540 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map