Assay Details
Target Gene Details
Entrez Gene ID: | 3586 |
Gene Name: | interleukin 10 |
Gene Aliases: |
CSIF, GVHDS, IL-10, IL10A, TGIF |
Location: |
Chr.1:206767603-206772494 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL10 | NM_000572.2 | NP_000563.1 | ||
| AF043333.1 | AAC03534.1 | |||
| AY029171.1 | AAK38162.1 | |||
| BC104252.1 | AAI04253.1 | |||
| BC104253.1 | AAI04254.1 | |||
| CR541993.1 | CAG46790.1 | |||
| CR542028.1 | CAG46825.1 | |||
| M57627.1 | AAA63207.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3578391 | Chr.1:206770892 - 206779577 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map