Assay Details
Target Gene Details
Entrez Gene ID: | 79085 |
Gene Name: | solute carrier family 25 member 23 |
Gene Aliases: |
APC2, MCSC2, SCaMC-3 |
Location: |
Chr.19:6436081-6460221 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2017710 | Chr.19:6441587 - 6442042 on Build GRCh38 | Deletion |
|
| nsv833727 | Chr.19:6400101 - 6568241 on Build GRCh38 | Loss |
|
| esv2718065 | Chr.19:6441562 - 6442041 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs188290935] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map