Assay Details
Target Gene Details
Entrez Gene ID: | 53832 |
Gene Name: | interleukin 20 receptor subunit alpha |
Gene Aliases: |
CRF2-8, IL-20R-alpha, IL-20R1, IL-20RA |
Location: |
Chr.6:136999971-137045366 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL20RA | NM_001278722.1 | 7 | 1676 | NP_001265651.1 |
| NM_001278723.1 | 6 | 1816 | NP_001265652.1 | |
| NM_001278724.1 | 6 | 1750 | NP_001265653.1 | |
| NM_014432.3 | 7 | 1995 | NP_055247.3 | |
| XM_006715506.2 | 6 | 1392 | XP_006715569.1 | |
| XM_011535904.2 | 6 | 1503 | XP_011534206.1 | |
| XM_017010954.1 | 7 | 2184 | XP_016866443.1 | |
| XM_017010955.1 | 7 | 1644 | XP_016866444.1 | |
| AF184971.1 | 7 | 1779 | AAF01320.1 | |
| AJ420490.1 | 3 | 970 | ||
| AK098312.1 | 3 | 1585 | ||
| AK297121.1 | 7 | 1638 | ||
| AK315293.1 | 7 | 1783 | ||
| AY358883.1 | 6 | 1330 | AAQ89242.1 | |
| BC113574.1 | 7 | 1873 | ||
| BC113602.1 | 7 | 1873 |
More Information
Set Membership: |
|

Genomic Map