Assay Details
Target Gene Details
Entrez Gene ID: | 4857 |
Gene Name: | NOVA alternative splicing regulator 1 |
Gene Aliases: |
Nova-1 |
Location: |
Chr.14:26445883-26597754 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NOVA1 | NM_002515.2 | 5 | 1201 | NP_002506.2 |
| NM_006489.2 | 4 | 1129 | NP_006480.2 | |
| XM_011536800.2 | 4 | 697 | XP_011535102.1 | |
| XM_017021342.1 | 5 | 1257 | XP_016876831.1 | |
| XM_017021343.1 | 4 | 1185 | XP_016876832.1 | |
| XM_017021344.1 | 6 | 1068 | XP_016876833.1 | |
| XM_017021345.1 | 5 | 1415 | XP_016876834.1 | |
| XM_017021346.1 | 6 | 1517 | XP_016876835.1 | |
| XM_017021347.1 | 6 | 1398 | XP_016876836.1 | |
| XM_017021348.1 | 5 | 1333 | XP_016876837.1 | |
| XM_017021349.1 | 4 | 1344 | XP_016876838.1 | |
| AK226114.1 | 6 | 1020 | ||
| AK291022.1 | 5 | 1024 | ||
| AK291468.1 | 5 | 902 | ||
| AK301526.1 | 6 | 985 | ||
| BC075038.2 | 5 | 906 | AAH75038.1 | |
| BC075039.2 | 5 | 907 | AAH75039.1 | |
| U04840.1 | 5 | 952 | AAA16022.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv564107 | Chr.14:26429752 - 26489004 on Build GRCh38 | Loss |
|
| nsv1052199 | Chr.14:26334093 - 26735737 on Build GRCh38 | Loss |
|
| esv3633850 | Chr.14:26411340 - 26535541 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map