Assay Details
Target Gene Details
Entrez Gene ID: | 5702 |
Gene Name: | proteasome 26S subunit, ATPase 3 |
Gene Aliases: |
TBP1 |
Location: |
Chr.11:47418769-47426473 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSMC3 | NM_002804.4 | NP_002795.2 | ||
| XM_017018026.1 | XP_016873515.1 | |||
| AK222485.1 | BAD96205.1 | |||
| AK291896.1 | ||||
| AK313518.1 | ||||
| BC008713.2 | ||||
| BC073165.1 | ||||
| BC106920.2 | AAI06921.1 | |||
| BC107804.1 | AAI07805.1 | |||
| BP284685.1 | ||||
| M34079.1 | AAA36666.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832141 | Chr.11:47311629 - 47499441 on Build GRCh38 | Loss |
|
| nsv554210 | Chr.11:47359042 - 47427993 on Build GRCh38 | Loss |
|
| nsv825871 | Chr.11:47354469 - 47435986 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map