Assay Details
Target Gene Details
Entrez Gene ID: | 25901 |
Gene Name: | coiled-coil domain containing 28A |
Gene Aliases: |
C6orf80, CCRL1AP |
Location: |
Chr.6:138773517-138793319 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC28A | NM_015439.2 | 1 | 54 | NP_056254.1 |
| XM_011535728.2 | 1 | 57 | XP_011534030.1 | |
| AY167571.1 | 1 | 72 | AAN87344.1 | |
| BC108717.1 | AAI08718.1 |
Target Gene Details
Entrez Gene ID: | 100507462 |
Gene Name: | GVQW motif containing 2 |
Gene Aliases: |
- |
Location: |
Chr.6:138725211-138773679 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GVQW2 | NM_001242740.1 | 1 | 107 | NP_001229669.1 |
| AK092592.1 | 1 | 80 | ||
| DN917422.1 | 1 | 107 |
More Information
Set Membership: |
|

Genomic Map