Assay Details
Target Gene Details
Entrez Gene ID: | 124 |
Gene Name: | alcohol dehydrogenase 1A (class I), alpha polypeptide |
Gene Aliases: |
ADH1 |
Location: |
Chr.4:99276366-99291028 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADH1A | NM_000667.3 | NP_000658.1 | ||
| AK290558.1 | ||||
| BC074738.2 | AAH74738.1 | |||
| BC117442.1 | ||||
| BC126306.1 | ||||
| BG199182.1 | ||||
| BT019812.1 | AAV38615.1 | |||
| M12271.1 | AAA68131.1 | |||
| M12963.1 | AAA51590.1 |
Target Gene Details
Entrez Gene ID: | 100507053 |
Gene Name: | uncharacterized LOC100507053 |
Gene Aliases: |
- |
Location: |
Chr.4:99088857-99301356 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100507053 | NR_037884.1 | |||
| BX647987.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5345n100 | Chr.4:99181544 - 99523288 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map