Assay Details
Target Gene Details
Entrez Gene ID: | 818 |
Gene Name: | calcium/calmodulin dependent protein kinase II gamma |
Gene Aliases: |
CAMK, CAMK-II, CAMKG |
Location: |
Chr.10:73812501-73874591 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAMK2G | NM_001204492.1 | 21 | 2452 | NP_001191421.1 |
| NM_001222.3 | 19 | 2320 | NP_001213.2 | |
| NM_172169.2 | 21 | 2416 | NP_751909.1 | |
| NM_172170.4 | 20 | 2389 | NP_751910.1 | |
| NM_172171.2 | 21 | 2503 | NP_751911.1 | |
| NM_172173.2 | 20 | 2347 | NP_751913.1 | |
| XM_005270195.1 | 22 | 2557 | XP_005270252.1 | |
| XM_005270197.1 | 22 | 2521 | XP_005270254.1 | |
| XM_005270198.1 | 22 | 2515 | XP_005270255.1 | |
| XM_005270199.1 | 21 | 2488 | XP_005270256.1 | |
| XM_005270200.1 | 21 | 2458 | XP_005270257.1 | |
| XM_005270201.1 | 21 | 2452 | XP_005270258.1 | |
| XM_005270203.1 | 21 | 2413 | XP_005270260.1 | |
| XM_005270205.1 | 20 | 2374 | XP_005270262.1 | |
| XM_006717993.1 | 23 | 2590 | XP_006718056.1 | |
| XM_006717996.1 | 22 | 2527 | XP_006718059.1 | |
| XM_006717997.1 | 22 | 2476 | XP_006718060.1 | |
| XM_006717998.1 | 22 | 2434 | XP_006718061.1 | |
| XM_017016728.1 | 21 | 2442 | XP_016872217.1 | |
| XM_017016730.1 | 21 | 2371 | XP_016872219.1 | |
| XM_017016733.1 | 20 | 2328 | XP_016872222.1 | |
| XM_017016735.1 | 20 | 2344 | XP_016872224.1 | |
| XM_017016736.1 | 21 | 2482 | XP_016872225.1 | |
| AK026309.1 | ||||
| AK094774.1 | ||||
| BC034044.1 | 21 | 2407 | AAH34044.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv46n21 | Chr.10:73790158 - 73933937 on Build GRCh38 | Loss |
|
| nsv551517 | Chr.10:73799109 - 73898823 on Build GRCh38 | Loss |
|
| nsv831912 | Chr.10:73649351 - 73840445 on Build GRCh38 | Gain |
|
| nsv1035634 | Chr.10:73724401 - 73907300 on Build GRCh38 | Gain |
|
| nsv825468 | Chr.10:73715689 - 73922563 on Build GRCh38 | Gain |
|
| nsv551516 | Chr.10:73749515 - 73847410 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map