Assay Details
Target Gene Details
Entrez Gene ID: | 119504 |
Gene Name: | anaphase promoting complex subunit 16 |
Gene Aliases: |
APC16, C10orf104, CENP-27, MSAG, bA570G20.3 |
Location: |
Chr.10:72216000-72235860 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANAPC16 | NM_001242546.1 | NP_001229475.1 | ||
| NM_001242547.1 | NP_001229476.1 | |||
| NM_001242548.1 | NP_001229477.1 | |||
| NM_173473.3 | NP_775744.1 | |||
| NR_038391.1 | ||||
| NR_038392.1 | ||||
| BC009530.1 | AAH09530.1 | |||
| BM768948.1 | ||||
| CB984695.1 | ||||
| CD511334.1 | ||||
| DA310509.1 | ||||
| DA739927.1 | ||||
| EU552924.1 |
More Information
Set Membership: |
|

Genomic Map