Assay Details
Target Gene Details
Entrez Gene ID: | 133383 |
Gene Name: | SET domain containing 9 |
Gene Aliases: |
C5orf35 |
Location: |
Chr.5:56909260-56928900 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SETD9 | NM_001171990.2 | 1 | 48 | NP_001165461.1 |
| NM_001323019.1 | 1 | 48 | NP_001309948.1 | |
| NM_001323022.1 | 1 | 48 | NP_001309951.1 | |
| NM_153706.3 | 1 | 48 | NP_714917.2 | |
| NR_136555.1 | 1 | 48 | ||
| NR_136556.1 | 1 | 48 | ||
| NR_136558.1 | 1 | 48 | ||
| XM_011543140.2 | XP_011541442.1 | |||
| AK309152.1 | ||||
| BC036528.1 | AAH36528.1 | |||
| BG719045.1 | 1 | 53 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv598210 | Chr.5:56881616 - 56958658 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map