Assay Details
Target Gene Details
Entrez Gene ID: | 10205 |
Gene Name: | myelin protein zero like 2 |
Gene Aliases: |
EVA, EVA1 |
Location: |
Chr.11:118253416-118264536 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MPZL2 | NM_005797.3 | NP_005788.1 | ||
| NM_144765.2 | NP_658911.1 | |||
| AF030455.1 | AAC39762.1 | |||
| AF275945.1 | AAF87240.1 | |||
| AF304447.1 | AAG23183.1 | |||
| AK290326.1 | ||||
| AY359061.1 | AAQ89420.1 | |||
| BC017774.1 | AAH17774.1 |
More Information
Set Membership: |
|

Genomic Map