Assay Details
Target Gene Details
Entrez Gene ID: | 401105 |
Gene Name: | uncharacterized LOC401105 |
Gene Aliases: |
AVPC1948 |
Location: |
Chr.3:188178543-188180808 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FLJ42393 | NR_024413.1 | 1 | 1179 | |
| AK124384.1 | 1 | 1179 | BAC85844.1 |
Target Gene Details
Entrez Gene ID: | 4026 |
Gene Name: | LIM domain containing preferred translocation partner in lipoma |
Gene Aliases: |
- |
Location: |
Chr.3:188153050-188890671 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LPP | NM_001167672.2 | NP_001161144.1 | ||
| XM_005247446.4 | XP_005247503.1 | |||
| XM_005247450.4 | XP_005247507.1 | |||
| XM_005247451.4 | XP_005247508.1 | |||
| XM_011512820.2 | XP_011511122.1 | |||
| XM_011512827.2 | XP_011511129.1 | |||
| XM_011512828.2 | XP_011511130.1 | |||
| XM_011512831.2 | XP_011511133.1 | |||
| XM_011512833.2 | XP_011511135.1 | |||
| XM_011512834.2 | XP_011511136.1 | |||
| XM_017006377.1 | XP_016861866.1 | |||
| XM_017006379.1 | XP_016861868.1 | |||
| XM_017006380.1 | XP_016861869.1 | |||
| AK304035.1 | ||||
| AL833171.1 | ||||
| BX474180.1 | ||||
| CD697098.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34193 | Chr.3:188129943 - 188550793 on Build GRCh38 | Loss |
|
| nsv1003486 | Chr.3:188157145 - 188284087 on Build GRCh38 | Gain |
|
| esv2751987 | Chr.3:188124933 - 188289510 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map