Assay Details
Target Gene Details
Entrez Gene ID: | 80020 |
Gene Name: | FAD dependent oxidoreductase domain containing 2 |
Gene Aliases: |
ERFAD |
Location: |
Chr.22:36487186-36507101 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FOXRED2 | NM_001102371.1 | 8 | 1827 | NP_001095841.1 |
| NM_024955.5 | 8 | 1866 | NP_079231.4 | |
| XM_017028941.1 | 7 | 1499 | XP_016884430.1 | |
| XM_017028942.1 | 7 | 1596 | XP_016884431.1 | |
| AK025714.1 | 8 | 1766 | BAB15227.1 | |
| AK026975.1 | 5 | 947 | BAB15610.1 | |
| AK091053.1 | 3 | 758 | ||
| AK225347.1 | 8 | 1766 | ||
| AK315555.1 | 8 | 1769 | ||
| BC015726.1 | 8 | 1750 | AAH15726.1 | |
| BC027716.2 | 8 | 1845 | AAH27716.1 | |
| BC040351.1 | 8 | 1789 | AAH40351.1 | |
| CR456444.1 | 7 | 1841 | CAG30330.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2760722 | Chr.22:36390921 - 36617980 on Build GRCh38 | Gain |
|
| nsv520497 | Chr.22:36299896 - 36570195 on Build GRCh38 | Gain |
|
| nsv1060004 | Chr.22:36448479 - 36615303 on Build GRCh38 | Gain |
|
| nsv834186 | Chr.22:36335878 - 36510765 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map