Assay Details
Target Gene Details
Entrez Gene ID: | 51204 |
Gene Name: | translational activator of cytochrome c oxidase I |
Gene Aliases: |
CCDC44 |
Location: |
Chr.17:63600872-63608365 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TACO1 | NM_016360.3 | 5 | 1199 | NP_057444.2 |
| AK094052.1 | 4 | 2035 | BAC04273.1 | |
| AL833861.1 | 3 | 612 | CAD38719.1 | |
| BC005049.1 | 5 | 1187 | AAH05049.1 | |
| BC007744.2 | 5 | 1011 | AAH07744.1 |
More Information
Set Membership: |
|

Genomic Map