Assay Details
Target Gene Details
Entrez Gene ID: | 11245 |
Gene Name: | G protein-coupled receptor 176 |
Gene Aliases: |
HB-954 |
Location: |
Chr.15:39795049-39920933 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR176 | NM_001271854.1 | 4 | 1402 | NP_001258783.1 |
| NM_001271855.1 | 3 | 563 | NP_001258784.1 | |
| NM_007223.2 | 3 | 1405 | NP_009154.1 | |
| XM_011521167.2 | 5 | 1053 | XP_011519469.1 | |
| XM_011521168.2 | 4 | 1544 | XP_011519470.1 | |
| XM_017021873.1 | 4 | 969 | XP_016877362.1 | |
| XM_017021874.1 | 4 | 1625 | XP_016877363.1 | |
| XM_017021875.1 | 5 | 1728 | XP_016877364.1 | |
| XM_017021876.1 | 5 | 1137 | XP_016877365.1 | |
| XM_017021877.1 | 7 | 1337 | XP_016877366.1 | |
| XM_017021878.1 | 2 | 1193 | XP_016877367.1 | |
| AK302715.1 | 3 | 563 | ||
| BC067106.1 | 4 | 735 | AAH67106.1 | |
| D38449.1 | 3 | 1405 | BAA07481.1 | |
| JQ308174.1 | 3 | 539 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832978 | Chr.15:39607558 - 39802680 on Build GRCh38 | Gain |
|
| esv3569465 | Chr.15:39178507 - 39912020 on Build GRCh38 | Gain |
|
| nsv832979 | Chr.15:39749727 - 39926206 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map