Assay Details
Target Gene Details
Entrez Gene ID: | 79695 |
Gene Name: | polypeptide N-acetylgalactosaminyltransferase 12 |
Gene Aliases: |
CRCS1, GalNAc-T12 |
Location: |
Chr.9:98807699-98850081 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALNT12 | NM_024642.4 | NP_078918.3 | ||
| XM_006717287.1 | XP_006717350.1 | |||
| XM_011519018.2 | XP_011517320.1 | |||
| XM_011519020.1 | XP_011517322.1 | |||
| XM_017015133.1 | XP_016870622.1 | |||
| AB040673.1 | BAD93346.1 | |||
| AB078146.1 | BAC07181.1 | |||
| AJ132365.2 | CAC80100.2 | |||
| AK024865.1 | BAB15027.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv614936 | Chr.9:98420416 - 98918098 on Build GRCh38 | Gain |
|
| esv3621214 | Chr.9:98651384 - 99637088 on Build GRCh38 | Gain |
|
| nsv825010 | Chr.9:98824981 - 98867887 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map