Assay Details
Target Gene Details
Entrez Gene ID: | 84795 |
Gene Name: | pyridine nucleotide-disulphide oxidoreductase domain 2 |
Gene Aliases: |
C10orf33, FP3420 |
Location: |
Chr.10:98383565-98415221 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PYROXD2 | NM_032709.2 | 12 | 1316 | NP_116098.2 |
| XM_011540293.2 | 13 | 2738 | XP_011538595.1 | |
| XM_011540301.2 | 5 | 2152 | XP_011538603.1 | |
| XM_017016835.1 | 12 | 1277 | XP_016872324.1 | |
| XM_017016836.1 | 12 | 1277 | XP_016872325.1 | |
| XM_017016837.1 | 12 | 1277 | XP_016872326.1 | |
| XM_017016838.1 | 12 | 3512 | XP_016872327.1 | |
| XM_017016839.1 | 10 | 1369 | XP_016872328.1 | |
| XM_017016840.1 | 10 | 1223 | XP_016872329.1 | |
| XM_017016841.1 | 9 | 1530 | XP_016872330.1 | |
| AK074429.1 | 12 | 1277 | ||
| AK075265.1 | 12 | 1254 | BAC11507.1 | |
| AK123713.1 | 2 | 1156 | ||
| AK225637.1 | 12 | 1270 | ||
| BC006131.1 | 12 | 1279 | AAH06131.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv515723 | Chr.10:98388419 - 98435072 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map