Assay Details
Target Gene Details
Entrez Gene ID: | 7532 |
Gene Name: | tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein gamma |
Gene Aliases: |
14-3-3GAMMA, PPP1R170 |
Location: |
Chr.7:76326791-76359025 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| YWHAG | NM_012479.3 | 2 | 462 | NP_036611.2 |
| AB024334.1 | 2 | 414 | BAA85184.1 | |
| AF142498.1 | 2 | 245 | AAD48408.1 | |
| AK024230.1 | 2 | 426 | ||
| AK293505.1 | ||||
| AK295029.1 | 3 | 341 | ||
| BC020963.2 | 2 | 436 | AAH20963.1 | |
| CR541904.1 | 2 | 245 | CAG46702.1 | |
| CR541925.1 | 2 | 245 | CAG46723.1 | |
| DC343104.1 | 2 | 490 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422494 | Chr.7:76286900 - 76984449 on Build GRCh38 | Duplication |
|
| nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain |
|
| nsv428174 | Chr.7:76305019 - 77220062 on Build GRCh38 | Gain |
|
| dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map