Assay Details
Target Gene Details
Entrez Gene ID: | 22843 |
Gene Name: | protein phosphatase, Mg2+/Mn2+ dependent 1E |
Gene Aliases: |
CaMKP-N, POPX1, PP2CH, caMKN |
Location: |
Chr.17:58755869-58985617 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPM1E | NM_014906.4 | 1 | 160 | NP_055721.3 |
| NR_048561.1 | 1 | 160 | ||
| AB028995.1 | 1 | 158 | ||
| AF260269.1 | 1 | 158 | AAF70325.1 | |
| AF520614.1 | AAM76058.1 | |||
| AK289966.1 | 1 | 160 | ||
| BC136290.1 | 1 | 100 | ||
| BC151228.1 | 1 | 158 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833498 | Chr.17:58652063 - 58815031 on Build GRCh38 | Loss |
|
| nsv1057312 | Chr.17:58748383 - 58804942 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map