Assay Details
Target Gene Details
Entrez Gene ID: | 57045 |
Gene Name: | twisted gastrulation BMP signaling modulator 1 |
Gene Aliases: |
TSG |
Location: |
Chr.18:9334767-9402420 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TWSG1 | NM_020648.5 | NP_065699.1 | ||
| AF196834.1 | AAG43468.1 | |||
| AF294628.1 | AAG10076.1 | |||
| AJ297391.1 | CAC05581.1 | |||
| AK027629.1 | BAB55246.1 | |||
| AK075550.1 | BAC11693.1 | |||
| AK315751.1 | ||||
| BC020490.1 | AAH20490.1 | |||
| DA735800.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056407 | Chr.18:9144347 - 9553766 on Build GRCh38 | Gain |
|
| nsv576447 | Chr.18:9240415 - 9357788 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map