Assay Details
Target Gene Details
Entrez Gene ID: | 122618 |
Gene Name: | phospholipase D family member 4 |
Gene Aliases: |
C14orf175 |
Location: |
Chr.14:104924806-104937789 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLD4 | NM_001308174.1 | NP_001295103.1 | ||
| NM_138790.3 | NP_620145.2 | |||
| XM_011536411.2 | XP_011534713.1 | |||
| XM_017020965.1 | XP_016876454.1 | |||
| AK295352.1 | ||||
| AK296188.1 | ||||
| AY358843.1 | AAQ89202.1 | |||
| BC015003.1 | AAH15003.2 | |||
| DC328186.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv566099 | Chr.14:104818822 - 104946204 on Build GRCh38 | Gain |
|
| nsv566139 | Chr.14:104880959 - 104950810 on Build GRCh38 | Loss |
|
| nsv1049253 | Chr.14:104918622 - 105005518 on Build GRCh38 | Gain |
|
| nsv952276 | Chr.14:104922864 - 105021963 on Build GRCh38 | Deletion |
|
| nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map