Assay Details
Target Gene Details
Entrez Gene ID: | 6881 |
Gene Name: | TATA-box binding protein associated factor 10 |
Gene Aliases: |
TAF2A, TAF2H, TAFII30 |
Location: |
Chr.11:6610817-6612244 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAF10 | NM_006284.3 | NP_006275.1 | ||
| AL833496.1 | 1 | 131 | ||
| BC012088.1 | AAH12088.1 | |||
| BM990572.1 | ||||
| CR541943.1 | CAG46741.1 | |||
| U13991.1 | AAA62230.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv553373 | Chr.11:6601484 - 6647097 on Build GRCh38 | Loss |
|
| nsv1035138 | Chr.11:6607812 - 6980555 on Build GRCh38 | Gain |
|
| esv996362 | Chr.11:6597567 - 6612421 on Build GRCh38 | Deletion |
|
| nsv825755 | Chr.11:6591998 - 6656613 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map