Assay Details
Target Gene Details
Entrez Gene ID: | 2595 |
Gene Name: | glucosidase alpha, neutral C |
Gene Aliases: |
- |
Location: |
Chr.15:42273658-42353666 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GANC | NM_001301409.1 | NP_001288338.1 | ||
| NM_001301410.1 | NP_001288339.1 | |||
| NM_198141.2 | NP_937784.2 | |||
| AK074330.1 | ||||
| BE893238.1 |
More Information
Set Membership: |
|

Genomic Map