Assay Details
Target Gene Details
Entrez Gene ID: | 140825 |
Gene Name: | neuralized E3 ubiquitin protein ligase 2 |
Gene Aliases: |
C20orf163, OZZ, OZZ-E3 |
Location: |
Chr.20:45888472-45891287 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NEURL2 | NM_001278535.1 | 2 | 1226 | NP_001265464.1 |
| NM_080749.3 | 2 | 1212 | NP_542787.1 | |
| AJ295985.1 | 2 | 917 | CAC82498.1 | |
| AK054821.1 | 2 | 1212 | BAB70810.1 | |
| AW298115.1 | 1 | 90 | ||
| AW515245.1 | 1 | 67 | ||
| BC074737.2 | AAH74737.1 | |||
| BC105935.1 | AAI05936.1 | |||
| BC107485.1 | AAI07486.1 | |||
| HY058356.1 | 1 | 235 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482069 | Chr.20:45888628 - 45891262 on Build GRCh38 | Gain |
|
| nsv517296 | Chr.20:45854586 - 45965936 on Build GRCh38 | Loss |
|
| nsv833991 | Chr.20:45885631 - 46068708 on Build GRCh38 | Loss |
|
| nsv833990 | Chr.20:45771510 - 45946844 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map