Assay Details
Target Gene Details
Entrez Gene ID: | 79828 |
Gene Name: | methyltransferase like 8 |
Gene Aliases: |
TIP |
Location: |
Chr.2:171315746-171434803 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
More Information
Set Membership: |
|

Genomic Map