Assay Details
Target Gene Details
Entrez Gene ID: | 337876 |
Gene Name: | chondroitin sulfate synthase 3 |
Gene Aliases: |
CHSY2, CSS3 |
Location: |
Chr.5:129903415-130186637 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHSY3 | NM_175856.4 | 1 | 128 | NP_787052.3 |
| XM_005271982.3 | 1 | 502 | XP_005272039.1 | |
| XM_005271983.3 | 1 | 502 | XP_005272040.1 | |
| XM_011543363.2 | XP_011541665.1 | |||
| XM_011543365.2 | 1 | 502 | XP_011541667.1 | |
| XM_017009435.1 | 1 | 502 | XP_016864924.1 | |
| AB086062.1 | 1 | 128 | BAC98832.1 | |
| AB175496.1 | 1 | 128 | ||
| AJ578034.2 | 1 | 486 | CAE17326.1 | |
| AJ632296.1 | 1 | 128 | CAG17593.1 | |
| AJ862728.1 | 1 | 128 | ||
| BC137325.1 | 1 | 197 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830488 | Chr.5:129843019 - 130014740 on Build GRCh38 | Gain |
|
| esv3890736 | Chr.5:129823851 - 130151212 on Build GRCh38 | Gain |
|
| esv2763498 | Chr.5:129423618 - 130021576 on Build GRCh38 | Gain |
|
| nsv599704 | Chr.5:129170896 - 130297608 on Build GRCh38 | Gain |
|
| nsv950389 | Chr.5:129904008 - 129906607 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map