Assay Details
Target Gene Details
Entrez Gene ID: | 55346 |
Gene Name: | t-complex 11 like 1 |
Gene Aliases: |
dJ85M6.3 |
Location: |
Chr.11:33039417-33079454 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TCP11L1 | NM_001145541.1 | NP_001139013.1 | ||
| NM_018393.3 | NP_060863.3 | |||
| XM_011520204.2 | XP_011518506.1 | |||
| XM_011520205.2 | XP_011518507.1 | |||
| XM_011520206.2 | XP_011518508.1 | |||
| XM_011520207.2 | XP_011518509.1 | |||
| XM_017017989.1 | XP_016873478.1 | |||
| XM_017017990.1 | XP_016873479.1 | |||
| XM_017017991.1 | XP_016873480.1 | |||
| XM_017017992.1 | XP_016873481.1 | |||
| AK002198.1 | BAA92131.1 | |||
| AK090646.1 | ||||
| BC041696.1 | AAH41696.1 | |||
| BP281502.1 | ||||
| DA340843.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1040484 | Chr.11:32929520 - 33071658 on Build GRCh38 | Gain |
|
| dgv1093n100 | Chr.11:33007427 - 33477800 on Build GRCh38 | Gain |
|
| esv2762904 | Chr.11:32947712 - 33564989 on Build GRCh38 | Gain |
|
| nsv521130 | Chr.11:33046843 - 33581367 on Build GRCh38 | Gain |
|
| nsv467786 | Chr.11:33043848 - 33455157 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map