Assay Details
Target Gene Details
Entrez Gene ID: | 84319 |
Gene Name: | cms1 ribosomal small subunit homolog (yeast) |
Gene Aliases: |
C3orf26 |
Location: |
Chr.3:99817834-100178632 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CMSS1 | NM_032359.3 | NP_115735.2 | ||
| AK124387.1 | ||||
| AK291392.1 | ||||
| AK312483.1 | ||||
| BC006475.1 | AAH06475.1 | |||
| BC006512.2 | AAH06512.1 |
Target Gene Details
Entrez Gene ID: | 11259 |
Gene Name: | filamin A interacting protein 1 like |
Gene Aliases: |
DOC-1, DOC1, GIP130, GIP90 |
Location: |
Chr.3:99833144-100114513 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FILIP1L | NM_001042459.2 | 5 | 3887 | NP_001035924.1 |
| NM_001282793.1 | NP_001269722.1 | |||
| NM_001282794.1 | NP_001269723.1 | |||
| NM_014890.3 | 2 | 2943 | NP_055705.2 | |
| NM_182909.3 | NP_878913.2 | |||
| XM_006713486.2 | 3 | 3139 | XP_006713549.1 | |
| XM_011512371.2 | 3 | 3005 | XP_011510673.1 | |
| AF329092.1 | AAN16206.1 | |||
| AF514867.1 | AAQ08177.1 | |||
| AF514868.1 | AAQ08178.1 | |||
| AF514869.1 | AAQ08179.1 | |||
| AK096195.1 | 3 | 2738 | ||
| AX776292.1 | CAE11726.1 | |||
| AX776294.1 | CAE11727.1 | |||
| AX776296.1 | CAE11728.1 | |||
| AX776298.1 | CAE11729.1 | |||
| AY642382.1 | AAV34207.1 | |||
| BC027860.1 | 2 | 2874 | AAH27860.1 | |
| BM674194.1 | 1 | 356 | ||
| EU531865.1 | ||||
| EU531866.1 | ||||
| U53445.1 | 2 | 2788 | AAA98972.1 |
More Information
Set Membership: |
|

Genomic Map