Assay Details
Target Gene Details
Entrez Gene ID: | 2833 |
Gene Name: | C-X-C motif chemokine receptor 3 |
Gene Aliases: |
CD182, CD183, CKR-L2, CMKAR3, GPR9, IP10-R, Mig-R, MigR |
Location: |
Chr.X:71615913-71618517 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CXCR3 | NM_001142797.1 | 2 | 486 | NP_001136269.1 |
| NM_001504.1 | 2 | 242 | NP_001495.1 | |
| XM_005262256.3 | 2 | 343 | XP_005262313.1 | |
| XM_005262257.3 | 2 | 343 | XP_005262314.1 | |
| XM_017029435.1 | 2 | 424 | XP_016884924.1 | |
| XM_017029436.1 | 2 | 240 | XP_016884925.1 | |
| AF469635.1 | 2 | 479 | AAP55851.1 | |
| AK313679.1 | 2 | 236 | ||
| AY242128.1 | 1 | 174 | AAO92295.1 | |
| BC034403.1 | 2 | 212 | AAH34403.1 | |
| KU178101.1 | 1 | 174 | ||
| KU178102.1 | 1 | 174 | ||
| X95876.1 | 2 | 242 | CAA65126.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6954 | Chr.X:71589852 - 71623669 on Build GRCh38 | Insertion |
|
| esv33082 | Chr.X:70420685 - 71888183 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map