Assay Details
Target Gene Details
Entrez Gene ID: | 5886 |
Gene Name: | RAD23 homolog A, nucleotide excision repair protein |
Gene Aliases: |
HHR23A, HR23A |
Location: |
Chr.19:12945814-12953643 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAD23A | NM_001270362.1 | 9 | 1506 | NP_001257291.1 |
| NM_001270363.1 | 8 | 1344 | NP_001257292.1 | |
| NM_005053.3 | 9 | 1509 | NP_005044.1 | |
| NR_072976.1 | 8 | 1453 | ||
| AI057203.1 | 1 | 325 | ||
| AI081136.1 | 1 | 328 | ||
| AK289908.1 | 9 | 1443 | ||
| AK293219.1 | ||||
| BC014026.2 | 9 | 1435 | AAH14026.1 | |
| BC088364.1 | 9 | 1435 | AAH88364.1 | |
| D21235.1 | 9 | 1410 | BAA04767.1 | |
| M77024.1 | 5 | 820 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953977 | Chr.19:12666887 - 13095486 on Build GRCh38 | Deletion |
|
| nsv833754 | Chr.19:12876208 - 13028186 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map