Assay Details
Target Gene Details
Entrez Gene ID: | 64412 |
Gene Name: | GDNF inducible zinc finger protein 1 |
Gene Aliases: |
ZBTB23, ZNF336 |
Location: |
Chr.20:23361585-23373062 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GZF1 | NM_001317012.1 | NP_001303941.1 | ||
| NM_001317019.1 | NP_001303948.1 | |||
| NM_022482.4 | NP_071927.1 | |||
| XM_005260800.2 | XP_005260857.1 | |||
| XM_005260802.3 | XP_005260859.1 | |||
| XM_011529321.2 | XP_011527623.1 | |||
| XM_011529322.1 | XP_011527624.1 | |||
| XM_017028017.1 | XP_016883506.1 | |||
| XM_017028018.1 | XP_016883507.1 | |||
| XM_017028019.1 | XP_016883508.1 | |||
| AB100265.1 | BAC98464.1 | |||
| AK025447.1 | BAB15134.1 | |||
| AK056159.1 | BAB71107.1 | |||
| AK056477.1 | ||||
| AK289814.1 | ||||
| AK293942.1 | ||||
| AK295362.1 | ||||
| AK314599.1 |
More Information
Set Membership: |
|

Genomic Map