Assay Details
Target Gene Details
Entrez Gene ID: | 84996 |
Gene Name: | URB1 antisense RNA 1 (head to head) |
Gene Aliases: |
C21orf119, PRED84 |
Location: |
Chr.21:32393133-32393960 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| URB1-AS1 | NR_026845.1 | 1 | 242 | |
| AK026376.1 | 1 | 220 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518458 | Chr.21:32393251 - 32402475 on Build GRCh38 | Loss |
|
| nsv834085 | Chr.21:32322748 - 32519107 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map