Assay Details
Target Gene Details
Entrez Gene ID: | 1010 |
Gene Name: | cadherin 12 |
Gene Aliases: |
CDHB |
Location: |
Chr.5:21750191-22853622 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDH12 | NM_001317227.1 | NP_001304156.1 | ||
| NM_001317228.1 | NP_001304157.1 | |||
| NM_004061.4 | NP_004052.2 | |||
| XM_011513927.2 | XP_011512229.1 | |||
| XM_017008920.1 | XP_016864409.1 | |||
| XM_017008921.1 | XP_016864410.1 | |||
| AK092176.1 | ||||
| AK295123.1 | ||||
| AK314800.1 | ||||
| BC047608.1 | AAH47608.1 | |||
| L33477.1 | AAB48539.1 | |||
| L34057.1 | AAA35623.1 |
More Information
Set Membership: |
|

Genomic Map