Assay Details
Target Gene Details
Entrez Gene ID: | 7030 |
Gene Name: | transcription factor binding to IGHM enhancer 3 |
Gene Aliases: |
RCCP2, RCCX1, TFEA, bHLHe33 |
Location: |
Chr.X:49028726-49043517 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TFE3 | NM_001282142.1 | NP_001269071.1 | ||
| NM_006521.5 | NP_006512.2 | |||
| AK295497.1 | ||||
| AK310095.1 | ||||
| AK313540.1 | ||||
| AL161985.1 | CAI46207.1 | |||
| BC026027.1 | AAH26027.1 | |||
| X51330.1 | CAA35714.1 | |||
| X96717.1 | CAA65478.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528292 | Chr.X:48433286 - 49274368 on Build GRCh38 | Gain |
|
| esv32909 | Chr.X:48003374 - 49285646 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map