Assay Details
Target Gene Details
Entrez Gene ID: | 56674 |
Gene Name: | TMEM9 domain family member B |
Gene Aliases: |
C11orf15 |
Location: |
Chr.11:8947201-8965011 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM9B | NM_001286094.1 | 4 | 1584 | NP_001273023.1 |
| NM_001286095.1 | 5 | 1233 | NP_001273024.1 | |
| NM_020644.2 | 5 | 1676 | NP_065695.1 | |
| AA433929.1 | ||||
| AB097017.1 | 5 | 1301 | BAC77370.1 | |
| AI346520.1 | 1 | 509 | ||
| AK074677.1 | 5 | 1294 | BAC11131.1 | |
| AK300712.1 | 5 | 1156 | ||
| AK316216.1 | 4 | 1215 | ||
| AY359069.1 | 5 | 1319 | AAQ89428.1 | |
| BC015884.1 | 4 | 1211 | AAH15884.1 | |
| BC040124.1 | 5 | 1358 | AAH40124.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv435923 | Chr.11:8817778 - 8979055 on Build GRCh38 | Deletion |
|
| esv2759803 | Chr.11:8763266 - 8988026 on Build GRCh38 | Gain |
|
| nsv832062 | Chr.11:8827748 - 8987925 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map